Xanthorrhizol 97

Supplier: ADIPOGEN CORP MS
Danger

AG-CN2-0090-M001
102987-314EA 276.92 USD
102987-314
Xanthorrhizol 97
Xanthorrhizol

Shows calcium-antagonistic and vasorelaxant activity. Antibacterial. Anti-inflammatory. Potent COX-2 and inducible nitric oxide (iNOS; NOS II) inhibitor. Hepatoprotective. Anticancer compound. Apoptosis inducer. Neuroprotective antioxidant. Antifungal.

Formula: C₁₅H₂₂O
MW: 218.3 g/mol
Storage Temperature: Freezer
CAS Number: 30199-26-9

Order Now

Specification Test Results

Source/Host Isolated from Curcuma xanthorrhiza.
Purity 97% (NMR)
Appearance/Color Colorless to light yellow oil.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR