Gag-Pol Retroviral Vector, Cell Biolabs
Supplier: Cell Biolabs
RV-111
102965-758EA 763.08 USD
102965-758
Gag-Pol Retroviral Vector, Cell Biolabs
Vectors, Plasmids and Libraries
pCMV-Gag-Pol Retroviral Vector contains both gag and pol genes from MMLV-based retrovirus
This vector may be co-transfected with other retroviral constructs to make recombinant retroviral particles.