Chrysomycin A Streptomyces species 98 HPLC_ASSAY_METHOD

Supplier: ADIPOGEN CORP MS

BVT-0099-C250 BVT-0099-M001
102513-548EA 193.85 USD
102513-548 102513-550
Chrysomycin A Streptomyces species 98 HPLC_ASSAY_METHOD
Chrysomycin A

Chrysomycin A is a Topoisomerase II inhibitor.


  • Antibiotic
  • Anti-neoplastic
  • Photosensitizing compound


Mediates a unique cross-linking reaction between DNA and histidine H3. Antitumor compound against human cell lines.


Solid


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Colour: Yellow to greenish

MW: 508.5 g/mol
Storage Temperature: Freezer
CAS Number: 82196-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR