Polyketomycin ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Antibiotic K99-5147, (2R,3S,6S)-2-methyl-6-{[(1S,3E,4aS,12aR)-4a,6,12a-trihydroxy-3-(1-hydroxyethylidene)-9-methoxy-11-methyl-2,4,5,7,10-pentaoxo-1,2,3,4,4a,5,7,10,12,12a-decahydrotetracen-1-yl]oxy}tetrahydro-2H-pyran-3-yl 2,6-dideoxy-4-O-(2-hydroxy-3,6-dimethylbenzoyl)-3-C-methyl-alpha-L-xylo-hexopyranoside

BVT-0033-M001
102513-478EA 193.85 USD
102513-478
Polyketomycin ≥98% (by HPLC)
Polyketomycin

  • Tetracyclic quinone glycoside
  • Antibacterial and antimalarial
  • Antitumor compound

Formula: C₄₄H₄₈O₁₈
MW: 864.8 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01938248
CAS Number: 200625-47-4

Order Now

Specification Test Results

Source/Host Isolated from Streptomyces sp.
Purity 98% (HPLC)
Appearance/Color Orange to dark brown solid.
Solubility Soluble in DMSO, methanol or acetone.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR