Wheaton Premium Services from DWK Life Sciences
Bar Coding & Taring
Customized bar code solutions to meet any application and taring services ensuring accuracy
DWK Life Sciences can customize a bar code solution to meet any application. It is critical that bar codes are read correctly every time, remain adhered to the container and are durable enough to withstand severe environments.
DWK Life Sciences premium services can provide you with pre-weighed containers meeting the highest expectations in convenience. Our taring services can ensure that the amount of material in the container will be accurately measured every time.
Convenience & Flexibility
- Increase productivity by freeing up time spent weighing & bar coding
- Sequence management ensures no duplicate bar codes
- Custom alpha/numeric bar code sequencing available to fit your application
- Choice symbologies: Code 39 and 128, 2D Matrix Code, QR Code, Interleaved 2 of 5
- Materials: Pressure sensitive, ceramic or direct mark
Critical Cleaning
Particulate cleaning to meet USP <788>. Depyrogenation with full documentation and certification. Sterilization to an acceptable Sterility Assurance Level (SAL)
Particulate Cleaning
DWK Life Sciences can deliver primary packaging components and labware particulate cleaned and validated to meet the allowable particulate limits according to USP <788>. DWK Life Sciences uses USP or WFI purified water, HEPA filtered and package items in certified class 100/10 cleanrooms.
- All products come with full documentation and certification
- Capable of processing glass and plastic products (closures &/ components)
- USP purified water rinse
- Capable of processing large or small volume containers
Depyrogenation
DWK Life Sciences depyrogenation techniques for glass containers and accessories include dry heat treatment and multiple WFI rinses to destroy and remove the pyrogens to endotoxin levels of less than 0.06 EU/mL with full documentation and certification.
- Receive ready-to-sterilize product
- Dry heat treatment or multiple WFI rinses
- Capable of processing glass and rubber products
- Capable of processing large and small volume containers
Sterilization
DWK Life Sciences can produce product to an acceptable Sterility Assurance Level (SAL). Typically a SAL of 10-6 is required; probability of a nonsterile unit is less than one in a million.
- All products come with full documentation and certification
- Receive ready-to-use product
- Single, double or triple autoclave bag options available
- Sterilization techniques for a range of different material compositions
- Sterile foil wrap process
- Small or large runs
Surface Treatment
A protective barrier between the contents and the glass component
Surface treatment refers to a process by which the entire inside surface of a container is deactivated via silanization or siliconization.
The coating provides a protective barrier between the contents and the glass, eliminating active sites on the glass that could potentially react with its contents.
DWK Life Sciences also has the ability to provide ammonium sulfate treated glass which protects against aqueous products that can react with alkali metal ions that are found in glass.
Convenience & Flexibility
- Minimize the interactions between the product and container
- Siliconization ensures higher restitution rate down to the last drop
- Silanization lubricates the vial and protects against alkalization