Volumetric Glassware
When your application demands precision and accuracy for dependable and repeatable results, turn to DWK Life Sciences — your partner on every lab journey. Our extensive portfolio of volumetric glassware includes the world’s most trusted brand names including, DURAN®, KIMBLE®, and KIMAX®. Designed, manufactured, and calibrated to deliver consistent accuracy, every piece meets or exceeds the most stringent ASTM and ISO regulatory requirements and our Class A, ASTM, and ISO glassware features thetightest tolerances available on the market.
Serialized and certified versions feature-complete traceability through a unique serial number printed on an accompanying certificate. And you can be confident that all calibrated glassware meets NIST certified standards.
Volumetric Flask
DWK offers the widest selection available on the market, including options from Class A, serialized and certified to economical Class B tolerances.
Volumetric Pipettes
DWK volumetric pipettes are manufactured according to both ASTM E543, and E969, are NIST traceable and will provide years of accurate service.
Graduated Cylinders
DWK volumetric cylinders are available in over 125 configurations from Class A serialized and certified to educational grade.
Burets
Over twenty design types, including educational grade and other specialized burets, meet broad application and budget needs.