Sterile Gloves
All Wells Lamont sterile glove liners are made and sterilized in the USA for high quality and quick shipping. These products have complete documentation and are trusted daily by end users in different applications.
Sterile low cut resistant cleanroom glove liner
Single use low level cut resistant Category III for particulates sterile glove liner.
Sterile moderate cut resistant cleanroom glove liner
ANSI A2 cut resistant reusable Category II for particulates sterile glove liner.
Sterile high cut resistant cleanroom glove liner
The M321 is a US made and sterilized glove liner that offers ANSI A4 cut resistance and tests Category III for particulates.