Mechanical Pipettes – Tacta®, mLINE® & Proline® Plus
Sartorius mechanical pipettes are appreciated for their ergonomics, ease of use and reliability. As a result, they are widely used for various liquid handling tasks in pharmaceutical laboratories, research institutes, universities, healthcare and industrial laboratories, where they help you to perform your manual pipetting easily and accurately. You may choose the pipette of your liking from the various models we offer to suite your specific needs and applications.
Tacta - Perfectly Balanced
Sartorius proudly presents Tacta, the new premium mechanical pipette with superb comfort and reliability. Tacta makes pipetting effortless and safe, while producing accurate and reliable results time after time. We listened to our customers and combined their views with our extensive knowledge and experienced R&D team to create the exceptionally well balanced design and performance of Tacta. Tacta feels great in your hand and is easy and comfortable to use. All materials used for Tacta have been carefully selected and each component is designed to meet the highest standards.
mLINE® – Our most advanced mechanical pipette
mLINE® – offers excellent ergonomics, performance and safety for manual pipetting. Its exceptionally low pipetting force and light-to-use tip ejection prevent repetitive strain injury (RSI) and ensure accurate results, also in long pipetting series. Increased safety with volume lock prevents accidental volume changes and unique Safe-Cone Filters minimize risk of contamination.
Proline Plus® – Dependable and Durable
Proline® Plus combines durable construction with ease and lightness of use, and is a perfect choice for liquid-handling professionals and students. The high chemical and UV resistance of Proline Plus will give you years of repeatable and accurate performance. Only three parts need to be disassembled for easy cleaning and maintenance. Fixed volume models are also available.