Multi Surface Flat Head Mops
Cleanroom Multi Surface Mops are well-suited for aseptic and controlled environment floors, walls, ceilings, windows, and other surfaces.
MZone™ FlatMop™ Multi-Surface Mop Covers
Slip-on, tab-removal flat head mop covers are well suited for cleaning and sanitizing all surfaces and equipment in the cleanroom. This mop easily glides onto adapters securely without any attachments.
SlideMop™ Flat Head Mop Covers
The SlideMop is a low touch mopping system with collapsible plates that easily slide into the quilted mop cover. Locks into place with its effortless step-on, downward-push system and releases with the click of a button.
SlimLine™ Flat Head Mop Covers
SlimLine™ Mop Covers easily and securely attach to the SlimLine™ Mop using elastic to snugly hold it in place. This mop is ideal for a variety of applications from disinfecting to quick clean ups.
SnapMop™ Flat Head Mop Covers™
SnapMop™ Covers are lightweight, snap-on mop covers with urethane foam interiors. They're designed for effective cleaning and sanitizing in the cleanroom. Snaps help secure cover for use with the LyoTool™ and Tank Tool™.
T-Mop™ Covered Sponge Mops
T-Mop™ Cleanroom Covers are rounded, covered sponges with snaps that fit over the 180° swivel T-Mop™ Adapter to easily roll over ceilings, down walls and soft walls as well as clean awkward corners, tight spaces, and under equipment.
Slim T™ Sponge Mops
Slim T™ Mop Covers are rounded, covered sponges that slip onto the T-Mop™ Adapter or Window Washing Tool to easily roll down walls and soft walls as well as clean awkward corners, tight spaces, and under equipment.
NovaSnap™ Flat Head Mop Covers
The NovaSnap™ low-touch flat mop head with a smooth knit polyester cleaning surface uses a simple snap-on and release clip action for improved cleanliness and ergonomics.