Liquid Sample Handling
We offer dispensing precision at your fingertips and organization for your workspace.
RF3000™ Pipet Controller
The RF3000™ accommodates glass and plastic pipets ranging in volume from 1 to 100 mL. The 0.45 μm nozzle filter prevents contamination and the 1 mL nozzle adapter helps keep small pipets stable. Five colored nose cones are included to help differentiate for departmental or specific-use coding. User labeling area on the unit allows for customization, instant recognition and/or task designation.
Pearl™ Adjustable Volume Single Channel Pipettors
The sleek ergonomic design and reduced plunger pressure help reduce the chance of RSI, as the pipettors require 2/3rd the operation pressure of most standard pipettors. All devices feature internal metal parts for increased accuracy and calibiration stability, easy volume adjustment with locking tab to prevent accidental change, a separate tip ejector and a large digital volume display.
Pipettor Workstation
Acrylic rack holds up to four pipettors on top shelf; dual bins house two different sizes of tips.
VWR® Universal Carousel Pipette Stand
The VWR® Universal Carousel Pipette Stand stores and holds pipettes in style. It holds up to eight single channel, four multi-channel, or a combination of both pipettes. A sleek and modern design enables this stand to securely hold virtually every known brand of pipette in the marketplace, making it truly ‘universal’.