Gas Chromatography
Gas Chromatography
Gas Chromatography (GC) is a technique for separating and analyzing compounds that can be vaporized without decomposition, and is widely used in environmental testing and forensic science, as well as for Pharma R&D and QC applications
Featured Suppliers:
Hamilton GC Autosampler Syringes
The C-Line syringe minimizes the potential for bent needles with a keyed flange design and ensures superior performance
Sampling
Sample Storage & Handling
Sample Preparation
Analysis
Results, Reviews & Data Storage
Featured Products
J.T. Baker Methanol ≥99.8%
BAKER ULTRA RESI-ANALYZED® Reagent—ultra high-purity solvents developed for organic residue extraction/concentration procedures. Suitable for exacting EPA sample cleanup protocols.
VWR® 20 mm Headspace Vials
Designed to fit most headspace autosamplers. Round bottom vials fit PerkinElmer, Tekmar, and Varian autosamplers. Flat bottom vials fit Agilent and Carlo Erba autosamplers.