FoodChek Actero Elite Enrichment Media
Speed and accuracy in food testing
The speed and accuracy of the patented Actero™ Elite Enrichment Media is your genuine food safety breakthrough solution, providing every service lab and food processor with increased efficiency and profitability while supporting brand protection, safe food products, and the health of humans and pets.
Actero™ Elite provides the ideal growing environment for injured or stressed contaminating Salmonella and Listeria, even in the presence of abundant competing flora, that allows for accelerated and reliable detection.
You, too, can experience the competitive advantages of Actero™ Elite through:
Single-step enrichment
- Reduce sample handling
- Use less enrichment media
Fastest "time-to-results"
- Improve workflow
- Higher throughput
Accurate test results
- Reduce food waste
- Release food products sooner
Actero™ is the brand to trust in delivering a reliable food safety solution!
Product selection
ACTERO™ Elite Salmonella Enrichment Media
Actero™ Elite Salmonella Enrichment Media received AOAC-PTM (041403) accreditation for the single-step recovery and enrichment of Salmonella spp.
ACTERO™ MediaBox
The Actero™ MediaBox™ is the solution for laboratories that require liquid culture media on-demand.
ACTERO™ Elite Listeria Enrichment Media
Actero™ Elite Listeria Enrichment Media received AOAC-PTM (111201) accreditation for the single-step recovery and enrichment of Listeria spp.
ACTERO™ EZ-Media Dry Bag
Actero™ EZ-Media Dry Bag increases safety and testing throughput, and reduces human error, time and cost in typical media preparation.