Controlled Environments
DuPont Cleanroom Apparel delivers an ideal balance of durability, comfort, and protection for use in critical environments within the biotechnology, pharmaceutical, microelectronics, semiconductor, food processing, and medical device manufacturing.
Sterile Cleanroom Manufacturing
Tyvek® IsoClean® Garments
- Comfortable, lightweight and durable
- Individually packaged garments offered as sterile or clean-processed & sterile
- Garments available gamma sterilized to an SAL of 10-6 with full traceability on all sterile apparel
- Serged or bound seam styles are available
Non-Sterile Cleanroom Manufacturing
Tyvek® IsoClean®, ProClean®, and ProShield® Garments
- Delivers the particle barrier, durability and comfort in use
- Available as individually packaged and clean processed as well as bulk packaged
- Variety of fabrics to meet your specific applications and needs
Controlled Environment and General Protection
Tyvek® IsoClean®, ProClean® and ProShield® Garments
- Delivers the particle barrier, durability and comfort in use
- Offers protection against non-hazardous particulates and light liquid splash
- Lightweight and low linting