Expert Consumable Manufacturing for Production Environments and Genomics
Through continuous new product innovation, Corning's® Axygen brand products offer new solutions for genomics sample handling, delivery and storage, improving the way research is done in the laboratory today. Axygen brand products include automation tips, pipet tips, PCR plates, deep well and storage plates, microtubes, sealing options, and the portfolio of nucleic acid purification kits.
Axygen® Automation Tips for Beckman Coulter i-Series, Biomek® Series, and Multimek™
A comprehensive line of tips for Beckman Coulter®
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are RNase-/DNase-/human gDNA-free and nonpyrogenic. Available in nonsterile, sterile, nonfiltered, filtered, wide bore, fine point, and extended length. Also available in Maxymum Recovery® surface to further improve accuracy and precision while reducing sample loss. Clear polypropylene tips are available in 96-well and 384-well format designed for Beckman Coulter AP-96, SPAN-8, MP20, MP200 and AP-384 head.
Genomics Solutions
From collection to analysis, we understand your challenges and your need for simplified, efficient, low- to high-throughput genomics processes.
Nucleic Acid Purification Kits