BRAND® Flexible Discharge Tubes for seripettor® Bottle-Top Dispensers
Supplier: Brandtech
704522 704523
12777-156EA 215.81 USD
12777-156 89042-170
BRAND® Flexible Discharge Tubes for seripettor® Bottle-Top Dispensers
Bottle Top Dispensers
Accessory parts for seripettor® and seripettor® pro bottle-top dispensers.
  • Includes adapter, sealing ring, and tube holder
  • PTFE tubing approximately 800 mm in length
  • Compatible with seripettor® and seripettor® pro
  • Not suitable with peroxides
The seripettor® and seripettor® pro are innovative bottle-top dispensers from BRAND®, designed for efficient and reliable dispensing of a wide range of polar solvents.
Optional accessories are designed for seamless compatibility with the seripettor® family of bottle-top dispensers. The flexible discharge tube speeds serial dispensing tasks, and permits fast and precise dispensing even into narrow test tubes or vials.