Brady® Eco-Friendly Matte White Polyester Labels for J7300 Printer
Supplier: Brady Worldwide
178389 178390 178391 178392 178393 178394 178395 178396 178397 178398 178388
77957-028RL 143.99 USD
77957-028 77957-114 77957-016 77957-040 77957-076 77957-088 77957-102 77957-082 77957-132 77957-106 77957-096
Brady® Eco-Friendly Matte White Polyester Labels for J7300 Printer
Labels
Boost sustainability without sacrificing performance. Inkjet receptive, eco-friendly polyester labels (B-2618) offer the durability of polyester with a reduced environmental impact.
  • Permanent acrylic adhesive
  • Service temperature: –40 to 100 °C (–40 to 212 °F)
Eco-friendly film contains recycled polyethylene terephthalate (PET). Ideal for rating plate applications and general-purpose labeling with high-quality barcodes and graphics. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction. Permanent acrylic adhesive ensures a lasting bond. Excellent resistance to most chemicals, oils, and water.
Ink cartridges are required to print on this material; use part numbers P-CMY-1 and P-K-1.
Certificaciones:
UL approved.