Brady® Eco-Friendly Matte White Polyester Labels for J4000 Printer
Supplier: Brady Worldwide
176668 176669 176670 176671 176672 176673 176674
77957-024RL 161.99 USD
77957-024 77957-128 77957-118 77957-086 77957-056 77957-116 77957-134
Brady® Eco-Friendly Matte White Polyester Labels for J4000 Printer
Labels
Boost sustainability without sacrificing performance. Inkjet receptive, eco-friendly polyester labels (B-2618) offer the durability of polyester with a reduced environmental impact.
  • Permanent acrylic adhesive
  • Service temperature: −40 to 212 °F (−40 to 100 °C)
Eco-friendly film contains recycled polyethylene terephthalate (PET). Ideal for rating plate applications and general-purpose labeling with high-quality barcodes and graphics. Permanent acrylic adhesive ensures a lasting bond. Excellent resistance to most chemicals, oils, and water.
An ink cartridge is required to print this material; use part number J40-CMY.
Certificaciones:
UL approved.