Nigericin sodium salt (from Streptomyces hygroscopicus)
Supplier: Chem-Impex International
Nigericin sodium salt, derived from Streptomyces hygroscopicus, is a potent ionophore known for its ability to facilitate the transport of potassium ions across cellular membranes. This unique property makes it invaluable in various research applications, particularly in studies involving ion transport mechanisms and cellular signaling pathways. Researchers utilize Nigericin sodium salt in pharmacological studies to explore its effects on cellular metabolism and its potential therapeutic applications in conditions such as hypertension and heart disease.
Moreover, Nigericin sodium salt is recognized for its role in enhancing the efficacy of certain antibiotics by disrupting the ionic balance within bacterial cells, thereby increasing their susceptibility to treatment. Its ability to modulate ion concentrations also makes it a useful tool in neurobiology, where it aids in understanding neurotransmitter release and neuronal excitability. With its diverse applications, Nigericin sodium salt stands out as a critical compound for researchers and industry professionals focused on advancing scientific knowledge and therapeutic strategies.
Formula:
C₄₀H₆₇O₁₁Na |
MDL Number:
MFCD30377206 CAS Number: 28380-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...