Static Mouse Cage, 1284L
Supplier: Tecniplast Usa
1284L001 1284L051 1284L116 1284L400SU 1284L400SUC 1284L016
10710-028CS 202.46 USD
10710-028 89042-318 77322-382 77322-384 77322-386 77969-670 77969-672 77969-668
Static Mouse Cage, 1284L
Animal Supplies Animal Cages
The 1284L Mouse Cage can comfortably house 4 to 5 mice of standard size, and is available in polycarbonate or polysulfone materials.
  • Made from transparent plastic with exceptional impact strength and heat resistance
  • 82 square inch cage base
  • Compatible with a variety of lids, tops, accessories, and racks
  • Stainless steel accessories
  • Can house 4 to 5 mice of standard size
  • Racks come in a variety of sizes and can come with or without protective bumpers
The Tecniplast 1284L conventional cage is designed for housing laboratory rodents, particularly mice, offering a standard size and configuration that ensures compatibility with other equipment in animal research facilities. It features high-quality, durable materials for long-lasting use and easy maintenance. The cage's smooth surfaces help reduce waste buildup, making cleaning more efficient while maintaining a hygienic environment for both animals and researchers.
With a focus on animal welfare, the 1284L cage provides a comfortable living space for the animals. Its design meets various animal welfare guidelines, offering enough space to reduce stress and enhance well-being. The cage is also designed for ease of handling, simplifying maintenance and monitoring and making it a practical choice for research applications.