VWR® Autoclavable Biohazard Bags, Polypropylene , Red
Supplier: VWR International
77778-318 77778-326 77778-332 77778-382 77778-392 77778-438 77778-340 77778-346 77778-430
77778-318CS 259.23 USD
77778-318 77778-326 77778-332 77778-382 77778-392 77778-438 77778-340 77778-346 77778-430
VWR® Autoclavable Biohazard Bags, Polypropylene , Red
Bags Sterilization Pouches
Sturdy, temperature-resistant bags are ideal for disposal of non-hazardous waste that must first be autoclaved. Bags can also be used for autoclaving other products.
  • Autoclavable up to 135 ℃ (275 ℉) maximum
  • Available in multiple thicknesses: 37.5 µm/1.5 mil or 50 µm/2 mil
  • Printed bags feature the universal biohazard symbol
  • Temperature indicator text ‘contents have been autoclaved’ turns dark to indicate successful autoclaving
  • Highly resistant to tears and punctures
Bags are supplied complete with 20 cm twist tie wire closures that allow expansion of the bag and ventilation when cooling. Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709.
Certificaciones:
ISO 9001:2015, Sedex.