Constant Climate Chambers, KBF
Supplier: Binder
9020-0498 9020-0499 9020-0494 9020-0495 9020-0496 9020-0497
77938-550EA 33187 USD
77938-550 77938-552 77938-542 77938-544 77938-546 77938-548
Constant Climate Chambers, KBF
Testing Chambers Climatic Chambers
The KBF series is a proven specialist for reliable ICH stability tests in the pharmaceutical industry: the KBF masters all long-term tests, accelerated tests and stress tests. It fulfills all important ICH requirements, from precision to programming and documentation. With flexible water supply options for all installation locations.
  • Energy-efficient cooling with modern inverter cooling unit
  • Cooling with climate-neutral R-600a refrigerant (GWP is 3)
  • Door heating to prevent condensation
  • Adjustable fan speed
  • Humidity regulation with capacitive humidity sensor and responsive steam humidification
  • Housing design with HIT technology
  • Inner doors made of metal oxide-coated safety glass
  • Large access port with silicone plug
  • Floor-standing units featuring robust twin casters with brakes at the front
  • Stackable tabletop units with a volume of up to 130 liters
  • Intuitive touchscreen controller with time-segment and real-time programming
  • Ergonomically adjustable control terminal (for units with a volume of 470 liters and above)
  • Signal triangle with light-up status display (for units with a volume of 470 liters and above)
  • Internal data logger, measured values can be read out in open format via USB
  • Class 3.3 independent temperature safety device (DIN 12880) with visual and acoustic alarms
  • Unit self-test for comprehensive status analysis
  • Data interfaces: Ethernet, USB
The units have a stainless-steel inner chamber and proven APT.line™ preheating chamber technology, feature a temperature range of 0 to 70 °C, and humidity range of 10 to 80% RH, the KBF consumes up to 40% less energy than its predecessor. With optional light modules, the KBF can be converted into an ICH photostability chamber and a plant growth chamber.
Información para pedidos:
Optional light modules for ICH photostability testing are available for 260, 470, 720 L models and optional LED light modules for plant growth are available for models from 260 L.
Información de suministro:
Supplied with 2 stainless steel racks and BINDER Multi Management Software APT-COM™ basic edition.