Gowning Mirrors
Supplier: PALBAM 2008 AC LTD TE
MR-01 MR-02 MR-03
77898-244EA 3366.45 USD
77898-244 77898-256 77898-290
Gowning Mirrors
Mirrors
Palbam Class is a world-renowned cleanroom gowning design and furniture supplier, with an extensive range of gownroom products and designs.
  • Cleanroom mirrors are available in three options
  • MR-01: Free standing, connected to the wall and floor
  • MR-02: Mounted in side panel of gown racks
  • MR-03: Mounted in side panel of bootie racks
Mirrors are an integral part of the gowning process. These cleanroom mirrors prioritize safety with an adhesive backing to prevent shattering. They are mounted on a stainless steel plate. The mirror and plate are then mounted
More Product Information