DNA/RNA Shield™ SafeCollect™ Swab Collection Kits
Supplier: Zymo Research
R1160 R1160-E R1161 R1161-E
76628-460EA 8.28 USD
76628-460 76628-462 76628-454 76628-456
DNA/RNA Shield™ SafeCollect™ Swab Collection Kits
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Sputum Sample Collection
The DNA/RNA Shield SafeCollect swab collection kit is a user-friendly collection kit for stabilizing the nucleic acid content of samples collected with a swab. DNA/RNA shield completely inactivates harmful pathogens in the sample, and stabilizes the RNA in a sample for up to 30 days at room temperature. The safe collect design eliminates the risk of spilling the stabilization reagent during use, and is ideal for applications that require at-home self collection.
  • User-friendly, spill-free design allows for safe, convenient collection of swab samples
  • Ideal for at-home self-collection applications
  • Applicable for diagnostics, COVID testing, NGS, qPCR
  • Completely inactivates harmful pathogens at the point of collection, for worry-free transport and downstream analysis
  • Stabilizes the DNA and RNA in a sample for transport/storage without the need for cold chain
  • Samples can be collected with a swab including buccal, nasal, throat, skin, surface, feces, etc