Saturix Sterile Isopropyl Alcohol
Supplier: FG CLEAN WIPES TE
6-LS7030VS-IPA-16B 6-LS7030VS-IPA-1G
77871-050CS 161.97 USD
77871-050 77871-052
Saturix Sterile Isopropyl Alcohol
Cleaning Agents Disinfectants
The dependability and performance of Saturix sterile alcohol spray is unmatched.
  • 70% USP grade Isopropyl Alcohol
  • 30% water for injection (WFI) quality
  • Alcohol blend filtered to 0.1 µm
  • Each bottle is double bagged
  • Pyrogen-free
  • Gamma irradiated and validated to a sterility level of 10⁻⁶
Fast-acting with extremely low chemical residue, this sterile IPA is manufactured in an ISO 6 cleanroom and packaged in an ISO 5 cleanroom, according to the highest GMP and ISO 9001 standards.