Accessory Shelf Kits for Ultra-Low Temperature Freezers
Supplier: Thermo Fisher Scientific
SK400TSX SK500TSX SK700TSX SK500TSX SK700TSX SK600TSX
10863-316EA 398 USD
10863-316 76019-594 76019-596 CA76019-594 CA76019-596 77681-942
Accessory Shelf Kits for Ultra-Low Temperature Freezers
Freezers Ultra Low Temperature Freezers
Organize your ULT freezer storage with additional interior stainless steel shelves.
  • Stainless steel shelf kit by freezer size
  • Includes shelf clips for installation
Certificaciones:
cULus and CE.
Envase:
One ULT shelf and installation clips provided.