TSX™ Core Series ULT Freezer Backup Systems
Supplier: Thermo Fisher Scientific
FCOPLC FCOTSXC FLNPLC FLNTSXC
77681-932EA 3479.9 USD
77681-932 77681-934 77681-936 77681-938
TSX™ Core Series ULT Freezer Backup Systems
Freezers Ultra Low Temperature Freezers
Keep precious samples protected with CO₂ and LN₂ backup systems for your ultra-low temperature freezers.
  • CO₂ Backup System: Maintains down to −67 °C
  • LN₂ Backup System: Maintains down to −86 °C
  • Includes backup battery explicitly used for the backup system
Certificaciones:
cULus and CE.