Human Recombinant CD40-Ligand (from E. coli)
Supplier: VWR International
77751-226 77751-228 77751-230 77751-232 77751-234
77751-226EA 434.3 USD
77751-226 77751-228 77751-230 77751-232 77751-234
Human Recombinant CD40-Ligand (from E. coli)
Proteins and Peptides
CD40-Ligand, or CD154, is a membrane glycoprotein and differentiation antigen that is expressed on the surface of T cells, monocytes, basophils, eosinophils, platelets, dendritic cells, and endothelial cells. CD40-Ligand binds and activates the CD40 receptor on antigen-presenting cells. CD40-Ligand stimulates B cell proliferation, immunoglobulin class switching, and antibody secretion.
  • High quality
  • Low endotoxin
  • Affordable
CD40-Ligand also induces cytokine production and tumoricidal activity in peripheral blood monocytes. CD40-Ligand is a co-stimulator of activated T cell proliferation and inflammatory protein production.
Precaución:
For research use only.