Horse Recombinant VEGF-165 (from E. coli)
Supplier: VWR International
77753-334 77753-336 77753-328 77753-330 77753-332
77753-334EA 3691.55 USD
77753-334 77753-336 77753-328 77753-330 77753-332
Horse Recombinant VEGF-165 (from E. coli)
Proteins and Peptides
Vascular endothelial growth factor-A (VEGF-A) is produced by a wide variety of cell types, including tumor and vascular cells. VEGF-A is a mediator of vascular growth, vascular permeability, and plays a role in stimulating vasodilation via nitric oxide-dependent pathways. VEGF-A has several alternatively spliced isoforms, with VEGF-165 being the most abundant. The VEGF-165 isoform is a secreted protein that acts on receptors VEGFR-1 and VEGFR-2 to modulate endothelial cell proliferation and angiogenesis.
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.