Rat Recombinant IFN-G (from E. coli)

Supplier: VWR International

77752-840 77752-842 77752-844 77752-846 77752-848
77752-840EA 239.88 USD
77752-840 77752-842 77752-844 77752-846 77752-848
Rat Recombinant IFN-G (from E. coli)
Proteins and Peptides

Interferon gamma (IFN-γ) is a type II interferon that is critical during adaptive and innate immune responses to infection.  IFN-γ is produced by T cells and natural killer cells following antigen-specific activation.  IFN-γ binds IFN-γ receptors (IFN-γ R1 and IFN-γ R2), which are expressed on most immune cells, to activate the JAK-STAT pathway.  IFN-γ-induced signaling increases the expression of class 1 major histocompatibility complex (MHC) molecules.  


  • High quality
  • Low endotoxin
  • Affordable


Precaución: For research use only.

New Product

Order Now

SPECIFICATIONS

Protein/Peptide Name IFN-G
Environmentally Preferable
Protein Synonyms Type II interferon|T cell interferon|Immune Interferon|MAF
Protein/Peptide Type Recombinant
Protein Function Cytokine
Species Rat
Source E. coli
Conjugation Unconjugated
Gene ID P01581
Sequence MQGTLIESLE SLKNYFNSSS MDAMEGKSLL LDIWRNWQKD GNTKILESQI ISFYLRLFEV LKDNQAISNN ISVIESHLIT NFFSNSKAKK DAFMSIAKFE VNNPQIQHKA VNELIRVIHQ LSPESSLRKR KRSRC
Protease-free N
Animal-Free N
Nuclease-free N
Carrier-Free Y
Endotoxin-free N
Endotoxin Level Low
Endotoxin Content Endotoxin LAL (EU/µg) ≤0.1
Biological Activity Bioactivity: No biological activity data is available at this time
Purity ≥95%
Grade RUO
Formulation 10 mM Sodium Phosphate, 100 mM Sodium Chloride, pH 7.5
Molecular Weight 15.6/31.2 kDa
Reconstitution Instructions Sterile water at 0.1 mg/ml
Storage Conditions –20 °C
Shipping Temperature RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR