Human Recombinant IL-6 (from E. coli)
Supplier: VWR International
77750-154 77750-156 77750-148 77750-150 77750-152
77750-154EA 2588.13 USD
77750-154 77750-156 77750-148 77750-150 77750-152
Human Recombinant IL-6 (from E. coli)
Proteins and Peptides
Interleukin 6 (IL-6) is an important pro-inflammatory and anti-inflammatory cytokine expressed by T cells, macrophages, and muscle cells.  IL-6 signals through a receptor complex containing two receptors, IL-6Rα and gp130.  IL-6 has an important function in promoting fever and can serve to stimulate an immune response to trauma.  IL-6 is often used for growth of hybridoma cell lines.  Human IL-6 is active on mouse and rat cells.  
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.