Human Recombinant EG-VEGF (from E. coli)
Supplier: VWR International
77750-098 77750-100 77750-102 77750-104 77750-106
77750-098EA 664.08 USD
77750-098 77750-100 77750-102 77750-104 77750-106
Human Recombinant EG-VEGF (from E. coli)
Proteins and Peptides
Endocrine gland-derived vascular endothelial growth factor (EG-VEGF) is an angiogenic growth factor that is expressed in the ovaries, testis, adrenal, and placental tissues. EG-VEGF has mitogenic, chemoattractive, and antiapoptotic functional roles. EG-VEGF signaling is mediated through binding the G protein-coupled receptors prokineticin receptor 1 (PKR1) and prokineticin receptor 2 (PKR2). Polycystic ovaries display strong EG-VEGF expression that is associated with increased angiogenesis and cyst formation, which could lead to the formation of polycystic ovary syndrome and infertility.
  • High quality
  • Low endotoxin
  • Affordable
Precaución:
For research use only.