Mouse Recombinant M-CSF (from E. coli)
Supplier: VWR International
77751-962 77751-964 77751-966 77751-968
77751-962EA 775.18 USD
77751-962 77751-964 77751-966 77751-968
Mouse Recombinant M-CSF (from E. coli)
Proteins and Peptides
Macrophage colony stimulating factor (M-CSF) is a hematopoietic growth factor that is widely produced by a variety of cells. M-CSF stimulates the proliferation and differentiation of hematopoietic stem cells into monocyte and macrophage cell types.  M-CSF also acts through the colony stimulating factor 1 receptor (CSF1R) to modulate processes involved in immunology, bone metabolism, fertility, and pregnancy.  Human M-CSF shows activity on mouse cells; however, mouse M-CSF shows no activity on human cells.  
  • High quality
  • Low endotoxin
  • Affordable
Precaución:
For research use only.