Human Recombinant IL-19 (from E. coli cells)
Supplier: VWR International
77751-834 77751-836 77751-838 77751-832
77751-834EA 262.6 USD
77751-834 77751-836 77751-838 77751-832
Human Recombinant IL-19 (from E. coli cells)
Proteins and Peptides
Interleukin-19 (IL-19) is a member of the interleukin 10 (IL-10) cytokine family and is produced by B cells and monocytes. IL-19 binds the interleukin 20 receptor complex (IL-20R) to activate STAT3 signaling. IL-19 induces interleukin 6 (IL-6) and tumor necrosis factor alpha (TNF-a) expression in monocytes, and promotes T helper 2 (Th2) cell-mediated immune responses.
  • High quality
  • Low endotoxin
  • Affordable
IL-19 production is up-regulated in resting monocytes following granulocyte-macrophage colony-stimulating factor (GM-CSF) or lipopolysaccharide (LPS) stimulation.
Precaución: For research use only.