Human Recombinant IL-4 (from E. coli)
Supplier: VWR International
77750-128 77750-130 77750-132 77750-134 77750-136
77750-128EA 623.68 USD
77750-128 77750-130 77750-132 77750-134 77750-136
Human Recombinant IL-4 (from E. coli)
Proteins and Peptides
Interleukin 4 (IL-4) is an immunomodulatory cytokine that functions to induce naïve helper T cells to differentiate into type 2 T helper (Th2) cells. Th2 cells subsequently produce more IL-4 in a positive feedback loop. IL-4 also promotes immunoglobulin IgG to IgE isotype switching on B cells. IL-4 binds the IL-4Rα receptor to activate STAT6 signaling.
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.