Human Recombinant M-CSF (from E. coli)
Supplier: VWR International
77753-398 77753-400 77753-402
77753-398EA 3047.68 USD
77753-398 77753-400 77753-402
Human Recombinant M-CSF (from E. coli)
Proteins and Peptides
Macrophage colony-stimulating factor (M-CSF) is a hematopoietic growth factor with broad production across different cell types (1). The regulation of monocyte and macrophage development to maintain homeostasis is primarily governed by M-CSF, which is generated by stromal cells present in both the bloodstream and tissues. Mature mononuclear phagocytes, in response, possess CSF-1 receptors (CSF-1R) and eliminate circulating CSF-1. This process creates a feedback mechanism responsible for diminishing monocyte proliferation (2, 3, 4). M-CSF also exerts its effects by interacting with CSF1R, influencing various processes related to immunology, bone health, fertility, and pregnancy regulation (5). Human M-CSF has cross reactivity to mouse cells, whereas mouse M-CSF does not exhibit activity on human cells (6).
  • High quality
  • Low endotoxin
  • Lot-to-lot consistency
  • Animal free
  • Manufactured following GMP guidelines
Host cell protein: ≤30 ng/ml, host cell DNA: ≤0.01 ng/ml, sterility testing: no growth, mycoplasma: negative.
Precaución: For research use or further manufacturing only.