Mouse Recombinant MCP-3 (from E. coli)
Supplier: VWR International
77752-354 77752-356 77752-358 77752-360
77752-354EA 600.95 USD
77752-354 77752-356 77752-358 77752-360
Mouse Recombinant MCP-3 (from E. coli)
Proteins and Peptides
Monocyte chemotactic protein 3 (MCP-3), also called CCL7, is produced by macrophages and tumor cell lines. MCP-3 signals through the G protein-coupled receptors CCR1, CCR2, and CCR3. MCP-3 chemoattracts monocytes and regulates macrophage function during inflammation and metastasis.
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.