Human Recombinant RH IL-2 C126S (from E. coli)
Supplier: VWR International
77753-338 77753-340 77753-342
77753-338EA 1004.95 USD
77753-338 77753-340 77753-342
Human Recombinant RH IL-2 C126S (from E. coli)
Proteins and Peptides
IL-2C126S is an immunomodulatory cytokine that is produced by lymphocytes. IL-2C126S signals through the IL-2R receptor to induce activated T cell proliferation and promote T cell differentiation. Stimulates the proliferation and differentiation of B cells, natural killer (NK) cells, monocytes, and macrophages. Promotes proliferation and differentiation of CAR T cells, T cells, and NK cells. Activates and expands T cells and NK cells from peripheral blood. The biological activity of CTGrade GMP interleukins and growth factors is standardized, where applicable, to WHO International standards, providing cell and gene therapy developers consistent, lot-to-lot biological activity and performance.
  • High quality
  • Low endotoxin
  • Lot-to-lot consistency
  • Animal free
  • Manufactured following GMP guidelines
Host cell protein: ≤30 ng/ml, host cell DNA: ≤0.01 ng/ml, sterility testing: no growth, mycoplasma: negative.
Precaución: For research use or further manufacturing only.