Human Recombinant PDGF-AA (from E. coli)
Supplier: VWR International
77750-674 77750-676 77750-670 77750-672
77750-674EA 6148.38 USD
77750-674 77750-676 77750-670 77750-672
Human Recombinant PDGF-AA (from E. coli)
Proteins and Peptides
Platelet-derived growth factor (PDGF) is an important regulator of cell growth, proliferation, and angiogenesis. PDGF synthesis is induced by IL-1, IL-6, TNF-α, TGF-β and EGF signaling. PDGF functions as a mitogenic growth hormone on cells of mesenchymal lineage, such as smooth muscle and glial cells. PDGF is also stored in the alpha-granules of platelets and is released upon adherence to traumatized tissues. PDGF is a dimeric glycoprotein formed by two A chains (AA), two B chains (BB), or as a heterodimer with an A and a B chain (AB). The PDGF dimer binds the cell surface receptor tyrosine kinases PDGFR-α and PDGFR-β.
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.