Mouse Recombinant GDNF (from E. coli)
Supplier: VWR International
77752-334 77752-328 77752-330 77752-332
77752-334EA 5065.15 USD
77752-334 77752-328 77752-330 77752-332
Mouse Recombinant GDNF (from E. coli)
Proteins and Peptides
Glial cell-derived neurotrophic factor (GDNF) is a neurotrophic factor that signals through a multicomponent receptor system to activate receptor tyrosine kinase RET signaling. GDNF promotes dopamine uptake, prevents motor neuron apoptosis, and supports the survival and differentiation of neurons.
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.