Mouse Recombinant GM-CSF (from E. coli )
Supplier:
VWR International
77752-058
77752-060
77752-062
77752-064
77752-066
77752-058EA
600.95
USD
77752-058
77752-060
77752-062
77752-064
77752-066
Mouse Recombinant GM-CSF (from E. coli )
Proteins and Peptides
Granulocyte-macrophage colony-stimulating factor (GM-CSF) is hematopoietic growth factor produced by endothelial cells, monocytes, fibroblasts, and T cells. GM-CSF stimulates the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells. GM-CSF promotes immune system development and regulates neutrophil function during infection. Human and mouse GM-CSF show no cross-reactivity.
- High quality
- Low endotoxin
- Affordable
Precaución: For research use only.