Mouse Recombinant IL-1 B (from E. coli)
Supplier: VWR International
77752-594 77752-596 77752-598 77752-592
77752-594EA 239.88 USD
77752-594 77752-596 77752-598 77752-592
Mouse Recombinant IL-1 B (from E. coli)
Proteins and Peptides
Interleukin 1 beta (IL-1β) is a pro-inflammatory cytokine that is produced by monocytes, macrophages, and dendritic cells (DCs). IL-1β signals through the IL-1 receptor, type 1 (IL-1R1) to activate the myeloid differentiation primary response 88 (MYD88) signaling pathway, which contains the cytoplasmic Toll/IL-1 receptor (TIR) domain adapter.
  • High quality
  • Low endotoxin
  • Affordable
IL-1β and the independently regulated IL-1α protein have overlapping proinflammatory activities to induce adhesion molecule expression on epithelial cells, control fever induction, initiate rheumatoid arthritis, and promote septic shock.
Precaución: For research use only.