Human Recombinant IL-5 (from E. coli)
Supplier: VWR International
77750-894 77750-896
77750-894EA 262.6 USD
77750-894 77750-896
Human Recombinant IL-5 (from E. coli)
Proteins and Peptides
Interleukin 5 (IL-5) is a hematopoietic growth factor that is expressed in type 2 T helper (Th2) cells, mast cells, and eosinophils. IL-5 acts through the IL-5 receptor (IL-5R), stimulates B cell growth, and mediates eosinophil activation. Human and mouse IL-5 show cross-reactivity.
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.