Mouse Recombinant IL-2 (from E. coli)
Supplier: VWR International
77752-114 77752-116 77752-108 77752-110 77752-112
77752-114EA 2638.63 USD
77752-114 77752-116 77752-108 77752-110 77752-112
Mouse Recombinant IL-2 (from E. coli)
Proteins and Peptides
Interleukin 2 (IL-2) is an immunomodulatory cytokine that is produced by lymphocytes. IL-2 signals through the IL-2R receptor to induce activated T cell proliferation and promote T cell differentiation. IL-2 also stimulates the proliferation and differentiation of B cells, natural killer cells, monocytes, and macrophages.
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.