Human Recombinant EBI3 (from E. coli)
Supplier: VWR International
77750-606 77750-608 77750-610 77750-614
77750-606EA 1015.05 USD
77750-606 77750-608 77750-610 77750-614
Human Recombinant EBI3 (from E. coli)
Proteins and Peptides
Epstein-Barr virus induced gene-3 (EBI3) is a secreted glycoprotein belonging to the hematopoietin receptor family related to the p40 subunit of interleukin 12 (IL-12). EBI3 expression is induced in B-lymphocytes in response to Epstein-Barr virus infection. EBI3 forms heterodimers with p28 to form interleukin 27 (IL-27), and with p35 to form interleukin 35 (IL-35). Both IL-27 and IL-35 have anti-inflammatory and regulatory activity.     
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.