Human Recombinant IL-7 (from E. coli)
Supplier: VWR International
77750-232 77750-234 77750-236 77750-238
77750-232EA 1015.05 USD
77750-232 77750-234 77750-236 77750-238
Human Recombinant IL-7 (from E. coli)
Proteins and Peptides
Interleukin 7 (IL-7) is a hematopoietic cytokine that is an important regulator of B and T cell development. IL-7 is secreted by bone marrow and thymic stromal cells, dendritic cells, intestinal epithelial cells, hepatocytes, and keratinocytes. IL-7 signals through the interleukin 7 receptor (IL-7R) to promote the differentiation of hematopoietic stem cells into T cells, B cells, and natural killer cells. IL-7 is also a regulator of intestinal mucosal lymphocyte proliferation. Human and mouse IL-7 show species cross-reactivity.
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.