VWR® Premium Touchscreen Laboratory Refrigerators
Supplier: VWR International
77696-000 77696-010 77696-002 77696-008 77696-012 77695-998
77696-000EA 14085.72 USD
77696-000 77696-010 77696-002 77696-008 77696-012 77695-998
VWR® Premium Touchscreen Laboratory Refrigerators
Refrigerators
The VWR® Premium Laboratory Touchscreen Refrigerators deliver superior cooling to laboratory environments.
  • Variable speed compressors
  • Premium touchscreen controllers
  • Microprocessor temperature control
  • Advanced alarm systems
  • Data logging capabilities
  • Natural hydrocarbon refrigerants
Engineered with variable speed compressors (VSCs), these units feature ultra-quiet operation and significant energy savings. VSCs optimize energy consumption by self-adjusting to cooling demands. These compressors also deliver enhanced system performance and provide a longer lifespan than other compressor variations.
The touchscreen line showcases the most elite features including state-of-the-art touchscreen controllers, advanced alarm systems and data logging capabilities. All units are powered by microprocessor temperature control. These units also utilize natural, hydrocarbon refrigerants for environmental health and energy efficiency. Upgrade your laboratory with these advanced units and welcome energy savings, noise reduction, smooth operation, and improved system performance to your work environment.
Certificaciones: ETL/cETL Listed