Anti-Varicella-Zoster Virus Human Recombinant Monoclonal Antibody [clone: RV37]

Supplier: ReVacc Scientific
N42Z57-100UG N42Z57-20UG
77761-704EA 997.02 USD
77761-704 77761-706
Anti-Varicella-Zoster Virus Human Recombinant Monoclonal Antibody [clone: RV37]
Antibodies

This antibody has been tested to bind to Varicella-Zoster virus (VZV) gE protein (Cat. No. B93H1). This antibody has been analyzed to compare its IgG format on non-denaturing gel.



Type: Primary
Antigen: VZV
Clonality: Recombinant monoclonaL
Clone: RV37
Conjugation:
Epitope:
Host: Human
Isotype: IgA1 Kappa
Reactivity: Human


Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR