Anti-hMPV Human Recombinant Monoclonal Antibody [clone: RV47]

Supplier: ReVacc Scientific
R7V27-100UG R7V27-20UG
77761-722EA 997.02 USD
77761-722 77761-724
Anti-hMPV Human Recombinant Monoclonal Antibody [clone: RV47]
Antibodies
This antibody has been tested to bind to fusion (F) glycoprotein of human Metapneumovirus (hMPV) subtype A1 strain (Cat. No. REVAM5Q48-A) or B1 strain (Cat. No. REVAM5Q48-B). An example of comparison of IgM and IgG using size exclusion chromatography is shown.

Type: Primary

Antigen: hMPV

Clonality: Recombinant Monoclonal

Clone: RV47

Conjugation: Unconjugated

Epitope:

Host: Human

Isotype: IgM kappa

Reactivity: Human


Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR