Anti-SARS and SARS-CoV-2 Human Recombinant Monoclonal Antibody [clone: RV88]

Supplier: ReVacc Scientific
P80H48-100UG P80H48-20UG
77761-714EA 997.02 USD
77761-714 77761-716
Anti-SARS and SARS-CoV-2 Human Recombinant Monoclonal Antibody [clone: RV88]
Antibodies
This antibody has been tested to bind to RBD of SARS (2003 strain) (Cat. No. 76515-456, 76515-458) and SARS-CoV-2 Wuhan Strain (Cat. No. 76515-436, 76515-438). An example of comparison of IgM and IgG using size exclusion chromatography is shown.

Type: Primary

Antigen: SARS and SARS-CoV-2

Clonality: Recombinant Monoclonal

Clone: RV88

Conjugation: Unconjugated

Epitope:

Host: Human

Isotype: IgM kappa

Reactivity: Human


Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR