Edge® 48-969 Medium-Duty Cut Protection Industrial Gloves, Ansell
Supplier: Ansell Healthcare
48969090 48969100 48969110 48969060 48969070 48969080
77867-848CS 1107.7 USD
77867-848 77867-850 77867-852 77867-842 77867-844 77867-846
Edge® 48-969 Medium-Duty Cut Protection Industrial Gloves, Ansell
Gloves
Economical solution for medium-duty applications requiring very high cut resistance, grip, and oil-repelling performance in oily environments.
  • NBR coated finish offers high grip in oily environments
  • Bright yellow design ensures high visibility
  • Thermal resistance allows the ability to confidently handle hot components
  • Superior cut-resistant lining for impact gloves with 360° cut resistance
  • Patented TPR design provides knuckle, thumb, and finger-length impact protection
Featuring a fully NBR-coated seamless liner with a rough palm, Edge® 48-969 protective gloves offer a highly secure grip in oily environments, while protecting skin from injuries and oil contamination. These bright yellow cut-resistant gloves ensure that hands remain highly visible, promoting safety even in low-light or dark conditions. The Edge® 48-969 gloves offer a high level of abrasion resistance and very high cut protection. In addition, the gloves provide EN 407 level 1 contact heat protection, making them ideal for workers handling hot equipment, parts, and objects.
Certificaciones:
ANSI/ISEA 105-2024 ABR Level 4|ANSI/ISEA 105-2024 Cut Level A7|ANSI/ISEA 105-2024 PUN Level 3|EN407:2020 |EN ISO 21420:2020|EN 388:2016 +A1:2018.
Envase:
Gloves are packaged 12 pairs/bag, 144 pairs/carton.